ContactUsLinkComponent
Reflex, Wound Clips, 7 mm, Roboz
Reflex, Wound Clips, 7 mm, Roboz
  100189-500
Reflex, Wound Clips, 7 mm, Roboz
  100189-500
 :  RS-9258
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Wound clips
  • Size:
    7 mm
  • Cat. No.:
    100189-500
  • Supplier no.:
    RS-9258
Reflex 7 mm Wound Clips; Stainless Steel; Box Of 100; For Use With RS-9250 Clip Applier.
These items are designed for animal research only and are not intended for human use.