Specifications
- Description:Molt Elevator, Double Ended
- Length:17.8 cm (7")
- Cat. No.:100190-152
- Supplier no.:RS-8830
Specifications
About this item
Molt Elevator; Double Ended; Both ends have blades with sharp edges.
One blade has a sharp, pointy tip while the other has a round tip.
Dimensions: Blade with sharp tip: 22m long x 4.5 mm wide x 1.5 mm thick. Blade with blunt tip: 25 mm long x 9 mm wide x 2.5 mm thick; 7" Length