Order Entry
Puerto Rico
ContactUsLinkComponent
Wecker Micro Dissecting Spatula, Roboz Surgical
Wecker Micro Dissecting Spatula, Roboz Surgical
  100190-138
Wecker Micro Dissecting Spatula, Roboz Surgical
  100190-138
 :  RS-6150

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Length:
    5"
  • Cat. No.:
    100190-138
  • Blade length:
    30 mm
  • Blade width:
    2 mm
  • Supplier no.:
    RS-6150

 

 

Wecker Micro Dissecting Spatula; 2 x 30 mm Blade; Sterling Silver; 5" Length