Order Entry
Puerto Rico
ContactUsLinkComponent
Forceps, 4" (10 cm) Long, Curved With Suture Tying Platform, 0.6 mm Tip, Roboz
Forceps, 4" (10 cm) Long, Curved With Suture Tying Platform, 0.6 mm Tip, Roboz
Catalog # 100189-666
Supplier:  Roboz Surgical
Forceps, 4" (10 cm) Long, Curved With Suture Tying Platform, 0.6 mm Tip, Roboz
Catalog # 100189-666
Supplier:  Roboz Surgical
Supplier Number:  RS-5097

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    100189-666
  • Supplier no.:
    RS-5097

Specifications

About this item

Harms Micro Suturing Forceps; Curved With Tying Platform; 0.6 mm Tip Width; 4" Length