ContactUsLinkComponent
Gemini Cautery System, Roboz Surgical
Gemini Cautery System, Roboz Surgical
Catalog # 100190-486
Supplier:  Roboz Surgical
Gemini Cautery System, Roboz Surgical
Catalog # 100190-486
Supplier:  Roboz Surgical
Supplier Number:  RS-300
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Gemini cautery system
  • Cat. No.:
    100190-486
  • Supplier no.:
    RS-300
Specifications
About this item
The gemini cautery System is the perfect tool for your cauterizing needs. The two tip sizes combined with two different operating temperatures allow for maximum versatility.
  • The tips are very easy to replace
  • Unit uses standard AA batteries
  • Handle has a screw-cap for convenient battery replacement
Using the battery spacer instead of one of the batteries changes the operating temperature from high to low.
Delivery: Kit contains a handle, two 5 mm tips, two 10 mm tips, two batteries and a battery spacer. The kit is shipped in hard plastic case with die cut foam for convenient storage. One year warranty available on this product.
Caution: These items are designed for animal research only and are not intended for human use.