ECL Western Blotting Substrate Kit, BioVision
Supplier: Avantor
Certificates
About this item
Superior Signal, clear background
- Convenient sizes (50 and 500 mL)
- Western blot analysis
BioVision ECL Western Blotting Substrate is a highly sensitive, nonradioactive, enhanced luminol-based chemiluminescent substrate for easy detection of horseradish peroxidase (HRP) on immunoblots. BioVision ECL Western Blotting Substrate offers superior signal and clear background. Blots can be repeatedly exposed to X-ray film or a CCD camera to obtain optimal results or stripped of the immunodetection reagents and re-probed. The special formulation of BioVision ECL substrate makes it ideal substitute for Amersham ECL substrate without the need for additional optimization of assay conditions.