Water USP, purified, sterile
Supplier: Cytiva
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Clear Liquid (Visual)
- Conductivity (Packaged):5.0 µs/cm (USP 645)
- Sterility:No Growth (USP 71)
- TOC (Before Packaging):500 ppb (USP 643)
Specification Test Results
Related Information
Certificates
Search by Lot NumberAT-Purified Water SDS
Datasheet
Cytiva BioXtra Buy-one-get-one US
Promotion Material
ghs_WPW-USP
SDS
RMBIO_Sterile_Water_and_Serum_Products_for_Academic_Research_Flyer