Anti-CSRNP2 Rabbit Polyclonal Antibody
Catalog # 89516-228
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Cysteine And Serine Rich Nuclear Protein 2
- Antigen Symbol:CSRNP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:207785
- Antigen Synonyms:member A1|C12orf2|MGC28165|family with sequence similarity 130|BC035295|cDNA sequence BC035295|C12orf22|TGF-beta induced apoptosis protein 12|TAIP-12 |FLJ25576
- Amino Acid Number:38 to 64
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:58.504 kDa
- Sequence:SILKRQKQLRRKC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This Mouse Csrnp2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 38-64 amino acids from the N-terminal region of mouse Csrnp2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89516-228
- Supplier no.:AP18600A
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: CSRNP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: