Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGB1 Mouse Monoclonal Antibody [clone: B-D15]
Anti-ITGB1 Mouse Monoclonal Antibody [clone: B-D15]
Catalog # 102995-488
Supplier:  Cell Sciences
Anti-ITGB1 Mouse Monoclonal Antibody [clone: B-D15]
Catalog # 102995-488
Supplier:  Cell Sciences
Supplier Number:  CDM034

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin beta 1
  • Antigen Symbol:
    ITGB1
  • Clonality:
    Monoclonal
  • Clone:
    B-D15
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    200 Tests
  • Antigen Synonyms:
    FNRB|integrin VLA-4 beta subunit|VLA-BETA|VLAB|VLA-4 subunit beta|CD29|beta polypeptide|MDF2|MSK12|GPIIA|Fibronectin receptor subunit beta|very late activation protein
  • Cat. No.:
    102995-488
  • Supplier no.:
    CDM034

Specifications

About this item

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: B-D15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human