Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD34 Mouse Monoclonal Antibody [clone: B-F23]
Anti-CD34 Mouse Monoclonal Antibody [clone: B-F23]
Catalog # 102995-406
Supplier:  Cell Sciences
Anti-CD34 Mouse Monoclonal Antibody [clone: B-F23]
Catalog # 102995-406
Supplier:  Cell Sciences
Supplier Number:  CDM179

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD34 Class III
  • Antigen Symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    B-F23
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    200 Tests
  • Cat. No.:
    102995-406
  • Supplier no.:
    CDM179

Specifications

About this item

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: B-F23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: