Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD34 Mouse Monoclonal Antibody [clone: B-F23]
  102995-402
Anti-CD34 Mouse Monoclonal Antibody [clone: B-F23]
  102995-402
 :  CDM177

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD34 Class III
  • Antigen Symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    B-F23
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    200 µg
  • Cat. No.:
    102995-402
  • Supplier no.:
    CDM177

 

 

Does not trigger KG-1a cell aggregation

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: B-F23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: