Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody [clone: B-E2]
Anti-CD2 Mouse Monoclonal Antibody [clone: B-E2]
  102995-468
Anti-CD2 Mouse Monoclonal Antibody [clone: B-E2]
  102995-468
 :  CDM024

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD2 molecule
  • Antigen Symbol:
    CD2
  • Clonality:
    Monoclonal
  • Clone:
    B-E2
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    200 µg
  • Antigen Synonyms:
    sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • Cat. No.:
    102995-468
  • Supplier no.:
    CDM024

 

 

Inhibits Mixed Lymphocyte Culture

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: B-E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human