Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD31 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B38]
  102995-492
Anti-CD31 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B38]
  102995-492
 :  CDM036

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD31 Molecule
  • Antigen Symbol:
    CD31
  • Clonality:
    Monoclonal
  • Clone:
    B-B38
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    endoCAM|PECAM-1|GPIIA'|EndoCAM|PECAM1|adhesion molecule|CD31|PECA1
  • Cat. No.:
    102995-492
  • Supplier no.:
    CDM036

 

 

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: B-B38
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: