Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRR5 Rabbit Polyclonal Antibody
Anti-PRR5 Rabbit Polyclonal Antibody
Catalog # 89515-106
Supplier:  Abgent
Anti-PRR5 Rabbit Polyclonal Antibody
Catalog # 89515-106
Supplier:  Abgent
Supplier Number:  AP19341B

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Proline Rich 5
  • Antigen Symbol:
    PRR5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    C-Terminal
  • Gene ID:
    109270
  • Antigen Synonyms:
    Protein Observed With Rictor-1|Protor 1|PROTOR1
  • Amino Acid Number:
    277 to 305
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    42.523 kDa
  • Sequence:
    EPQGFVDTPGQGPSC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This Mouse Prr5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 277-305 amino acids from the C-terminal region of mouse Prr5.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89515-106
  • Supplier no.:
    AP19341B

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: PRR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse