Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-E3]
Anti-CD10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-E3]
Catalog # 102995-374
Supplier:  Cell Sciences
Anti-CD10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-E3]
Catalog # 102995-374
Supplier:  Cell Sciences
Supplier Number:  CDM144

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Neprilysin
  • Antigen Symbol:
    CD10
  • Clonality:
    Monoclonal
  • Clone:
    B-E3
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    Calla|Common Acute Lymphoblastic Leukemia Antigen|Nep|Mme|Cd10|Membrane Metallo-Endopeptidase|Cluster Of Differentiation 10|Neutral Endopeptidase
  • Cat. No.:
    102995-374
  • Supplier no.:
    CDM144

Specifications

About this item

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: B-E3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human