Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody (Biotin) [clone: B-B8]
Anti-CD5 Mouse Monoclonal Antibody (Biotin) [clone: B-B8]
  102995-366
Anti-CD5 Mouse Monoclonal Antibody (Biotin) [clone: B-B8]
  102995-366
 :  CDM134

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD5 molecule
  • Antigen Symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    B-B8
  • Conjugation:
    Biotin
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • Cat. No.:
    102995-366
  • Supplier no.:
    CDM134

 

 

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: B-B8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: