Anti-SPATA19 Rabbit Polyclonal Antibody
89512-868
: Abgent
- Antibody Type:Primary
- Antigen Name:Spermatogenesis Associated 19
- Antigen Symbol:SPATA19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:219938
- Antigen Synonyms:spergen1|CT132|SPAS1
- Amino Acid Number:122 to 150
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:19.186 kDa
- Sequence:RDRIEQVRRSISRLC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This SPT19 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 122-150 amino acids from the C-terminal region of human SPT19.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89512-868
- Supplier no.:AP13095B
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,
Type: Primary
Antigen: SPATA19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse