Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody [clone: B-B8]
Anti-CD5 Mouse Monoclonal Antibody [clone: B-B8]
Catalog # 102995-362
Supplier:  Cell Sciences
Anti-CD5 Mouse Monoclonal Antibody [clone: B-B8]
Catalog # 102995-362
Supplier:  Cell Sciences
Supplier Number:  CDM132

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD5 molecule
  • Antigen Symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    B-B8
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    200 µg
  • Antigen Synonyms:
    CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • Cat. No.:
    102995-362
  • Supplier no.:
    CDM132

Specifications

About this item

Immunohistochemistry (recommended dilution 1:50)

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: B-B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: