Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD11A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B15]
Anti-CD11A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B15]
Catalog # 102995-476
Supplier:  Cell Sciences
Anti-CD11A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B15]
Catalog # 102995-476
Supplier:  Cell Sciences
Supplier Number:  CDM028

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Alpha L
  • Antigen Symbol:
    CD11A
  • Clonality:
    Monoclonal
  • Clone:
    B-B15
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    LFA-1 alpha|LFA-1|Leukocyte adhesion glycoprotein LFA-1 alpha chain|antigen CD11A (p180)|LFA1A|Leukocyte function-associated molecule 1 alpha chain|CD11A|alpha polypeptide|CD11 antigen-like family member A|LFA-1A|lymphocyte function-associated antigen 1
  • Cat. No.:
    102995-476
  • Supplier no.:
    CDM028

Specifications

About this item

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: B-B15
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human