Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD130 Mouse Monoclonal Antibody [clone: B-S12]
Catalog # 102995-458
Supplier:  Cell Sciences
Anti-CD130 Mouse Monoclonal Antibody [clone: B-S12]
Catalog # 102995-458
Supplier:  Cell Sciences
Supplier Number:  CDM019

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Interleukin 6 Signal Transducer
  • Antigen Symbol:
    IL6ST
  • Clonality:
    Monoclonal
  • Clone:
    B-S12
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    200 µg
  • Antigen Synonyms:
    IL-6RB|CD130 antigen|IL-6 receptor subunit beta|GP130|gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form|oncostati|IL-6ST|IL-6R subunit beta|CDW130|membrane glycoprotein 130|membrane glycoprotein gp130|interleukin receptor beta chain
  • Cat. No.:
    102995-458
  • Supplier no.:
    CDM019

Specifications

About this item

Induces gp130 activation, Jak 1, Jak 2, Stat 1 and Stat 3 phosphorylation. Activates cells carrying gp130

Type: Primary
Antigen: CD130
Clonality: Monoclonal
Clone: B-S12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human