Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:HLA Class II (DR)
- Clonality:Monoclonal
- Clone:B-C10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Size:200 Tests
- Cat. No.:102995-424
- Supplier no.:CDM347
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.
Type: Primary
Antigen: HLA Class II (DR)
Clonality: Monoclonal
Clone: B-C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: