Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:IgG Fc Receptor 3
- Antigen Symbol:IGFR3
- Clonality:Monoclonal
- Clone:B-E16
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 Tests
- Antigen Synonyms:CD16|FCRIII|CD16A|FCGR3|IMD20|FCG3|FCRIIIA|FCR-10|FCGRIII|IGFR3
- Cat. No.:102995-382
- Supplier no.:CDM154
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: B-E16
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human