Pinnacle® DB Silica HPLC Columns (USP L3), Restek
: Restek
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.
- For normal phase separations
- Optimized for UHPLC stability
Pinnacle DB columns are 100% manufactured by Restek in our Bellefonte, Pennsylvania facility. Because performance begins with the support, our Pinnacle DB UHPLC columns start with base-deactivated spherical silica that is optimized for UHPLC stability. From there, we bond them with a wide variety of phases to give chromatographers a stable and selective column. Get the most out of your UHPLC system. Combine selectivity and efficiency by using Restek Pinnacle DB UHPLC columns.
- Catalog No:
- 10841-030
- 10838-522
- 10837-608
- 10837-604
- 10837-602
- 10837-598
- 10059-836
- 10837-614
- 10059-834
- 10059-832
- 10837-610
- 10838-506
- 10838-514
- 10841-028
- 10838-498
- 10837-664
- 10837-662
- 10838-482
- 10838-490
- 10837-652
- 10837-650
- 10837-646
- 10837-644
- 10837-640
- 10837-638
- 10838-476
- 10838-466
- 10838-474
- 10838-458
- 10837-628
- 10837-626
- 10837-622
- 10837-620
- 10837-616
- 10839-736
- pH Range:
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- 2.5 - 8.0
- Temperature Limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP Code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3