Ultra Cyano HPLC Prep Columns, Restek
: Restek
The Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired.
- Particle size and pore size: 5 µm, spherical and 100 Å
- Carbon load: 8%
- End-cap: yes
- pH range: 2.5 to 8
- Temperature limit: 80 °C
- USP phase code: L10
- Phase category: cyano
- Ligand type: cyanopropylsilane
It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases. Easy scale-up from Restek analytical columns.