Graether Collar Button, OR Grade, Sklar
Product Details & Documents
For iris retraction. Constructed of stainless steel.
- Collar Button 45° Angle
- 10mm from Bend to Tip
Industry standard.
Specifications
- Description:Button End Graether Collar Button
- Curvature:Angled (45°)