ContactUsLinkComponent
Cannula with Tip Only for Universal Style Biopsy Punch, OR Grade, Sklar
Cannula with Tip Only for Universal Style Biopsy Punch, OR Grade, Sklar
Catalog # 10198-890
Supplier:  Sklar
undefined
Cannula with Tip Only for Universal Style Biopsy Punch, OR Grade, Sklar
Catalog # 10198-890
Supplier:  Sklar
Supplier Number:  90-7557
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mini Tischler Universal Style Biopsy Punch Canula and Tip
  • Cat. No.:
    10198-890
  • Supplier no.:
    90-7557
Specifications
About this item
Sklar's Mini Tischler Universal Style Biopsy Punch Canula and Tip is part of an OB/GYN instrument
This product attaches to a universal style handle and together the complete instrument they create are biopsy punch forceps. They are often used in colposcopy procedures to obtain cervical tissue samples for biopsy. The mini Tischler bite, 2.3x4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because less bleeding typically occurs when the smaller tissue sample is removed.