Membrane Filter Discs, PTFE, Hydrophobic, Advantec
About this item
Advantec offers both supported and unsupported Hydrophobic PTFE membrane filters.
Grade J-type is laminated with a polypropylene mesh support which offers improved handling. Operating temperature range: –35 to 130 °C.
Grade T-type is unsupported. Operating temerature range is –120 to 260 °C.
Applications: Sterilize gases and trap aqueous aerosols. Protect vacuum pumps by allowing gases to pass freely while blocking aqueous liquids. Vent critical samples. Sterilize and clarify strong acids and many other solvents incompatible with other membranes.
This filter media is inherently hydophobic. The filter is thin, highly porous and behaves as an absolute retentive membrane. Inert to most chemically aggressive solvents, strong acids and bases, PTFE membranes offer maximum chemical and pH resistance.
66 Options Available Below
Product Details & Documents
Advantec offers both supported and unsupported Hydrophobic PTFE membrane filters.
Grade J-type is laminated with a polypropylene mesh support which offers improved handling. Operating temperature range: –35 to 130 °C.
Grade T-type is unsupported. Operating temerature range is –120 to 260 °C.
Applications: Sterilize gases and trap aqueous aerosols. Protect vacuum pumps by allowing gases to pass freely while blocking aqueous liquids. Vent critical samples. Sterilize and clarify strong acids and many other solvents incompatible with other membranes.
This filter media is inherently hydophobic. The filter is thin, highly porous and behaves as an absolute retentive membrane. Inert to most chemically aggressive solvents, strong acids and bases, PTFE membranes offer maximum chemical and pH resistance.
Documents
Certificates
Search by Lot NumberProduct Family Options
Supported J-Type
Product Information
- Pore sizeØPorosityThicknessFlow rateAvailabilityPrice
- Supplier Number: J010A013ASpecifications:MarketsourceMarketsource
- 0.10 µm25 mm72%130 µm39.1 mL/min/cm2 (Acetone)Log in for availabilityPack of 100 (1/Itm/Bx)$335.95Supplier Number: J010A025ASpecifications:MarketsourceMarketsource
- 0.10 µm47 mm72%130 µm39.1 mL/min/cm2 (Acetone)Log in for availabilityPack of 100 (1/Itm/Bx)$450.19Supplier Number: J010A047ASpecifications:MarketsourceMarketsource
- Supplier Number: J010A090CSpecifications:MarketsourceMarketsource
- 0.10 µm142 mm72%130 µm39.1 mL/min/cm2 (Acetone)Log in for availabilityPack of 25 (1/Itm/Bx)$577.32Supplier Number: J010A142CSpecifications:MarketsourceMarketsource
- 0.10 µm293 mm72%130 µm39.1 mL/min/cm2 (Acetone)Log in for availabilityPack of 10 (1/Itm/Bx)$780.02Supplier Number: J010A293DSpecifications:MarketsourceMarketsource
- Supplier Number: J020A013ASpecifications:MarketsourceMarketsource
- Supplier Number: J020A025ASpecifications:MarketsourceMarketsource
- Supplier Number: J020A037ASpecifications:MarketsourceMarketsource
- Supplier Number: J020A047ASpecifications:MarketsourceMarketsource
- Supplier Number: J020A050ASpecifications:MarketsourceMarketsource
- Supplier Number: J020A090CSpecifications:MarketsourceMarketsource
- Supplier Number: J020A142CSpecifications:MarketsourceMarketsource
- 0.20 µm293 mm72%130 µm61.4 mL/min/cm2 (Acetone)Log in for availabilityPack of 10 (1/Itm/Bx)$780.02Supplier Number: J020A293DSpecifications:MarketsourceMarketsource
- Supplier Number: J050A013ASpecifications:MarketsourceMarketsource
- Supplier Number: J050A025ASpecifications:MarketsourceMarketsource
- Supplier Number: J050A037ASpecifications:MarketsourceMarketsource
- Supplier Number: J050A047ASpecifications:MarketsourceMarketsource
- Supplier Number: J050A050ASpecifications:MarketsourceMarketsource
- Supplier Number: J050A090CSpecifications:MarketsourceMarketsource
- Supplier Number: J050A142CSpecifications:MarketsourceMarketsource
- Supplier Number: J050A293DSpecifications:MarketsourceMarketsource
- Supplier Number: J100A013ASpecifications:MarketsourceMarketsource
- Supplier Number: J100A025ASpecifications:MarketsourceMarketsource
- Supplier Number: J100A037ASpecifications:MarketsourceMarketsource
- Supplier Number: J100A047ASpecifications:MarketsourceMarketsource
- Supplier Number: J100A050ASpecifications:MarketsourceMarketsource
- Supplier Number: J100A090CSpecifications:MarketsourceMarketsource
- Supplier Number: J100A142CSpecifications:MarketsourceMarketsource
- Supplier Number: J100A293DSpecifications:MarketsourceMarketsource
Unsupported T-Type
Product Information
- Pore sizeØPorosityThicknessFlow rateAvailabilityPrice
- Supplier Number: T010A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T010A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T010A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T010A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T010A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T010A293DSpecifications:MarketsourceMarketsource
- Supplier Number: T020A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T020A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T020A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T020A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T020A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T020A293DSpecifications:MarketsourceMarketsource
- Supplier Number: T050A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T050A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T050A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T050A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T050A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T050A293DSpecifications:MarketsourceMarketsource
- Supplier Number: T080A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T080A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T080A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T080A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T080A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T080A293DSpecifications:MarketsourceMarketsource
- Supplier Number: T100A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T100A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T100A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T100A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T100A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T100A293DSpecifications:MarketsourceMarketsource
- Supplier Number: T300A013ASpecifications:MarketsourceMarketsource
- Supplier Number: T300A025ASpecifications:MarketsourceMarketsource
- Supplier Number: T300A047ASpecifications:MarketsourceMarketsource
- Supplier Number: T300A090CSpecifications:MarketsourceMarketsource
- Supplier Number: T300A142CSpecifications:MarketsourceMarketsource
- Supplier Number: T300A293DSpecifications:MarketsourceMarketsource



