Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Neuron Speci Enolase (from E. coli)
Human Recombinant Neuron Speci Enolase (from E. coli)
Catalog # 10008-416
Supplier:  Avantor
CAS Number:  
undefined
Human Recombinant Neuron Speci Enolase (from E. coli)
Catalog # 10008-416
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 μg
  • Endotoxin Content:
    <1.0 EU per 1 μg of protein (determined by LAL method).
  • Enzyme Name:
    Neuron Speci Enolase
  • Enzyme Activity:
    Specific activity: >8 units/ml. Size: 100ug
  • Enzyme Synonyms:
    Enolase 2, gamma, neuronal, ENO2
  • Purity:
    ≥95%
  • Molecular Weight:
    47 kDa
  • Formulation:
    1 mg/ml solution in 20 mM Tris-HCl (pH 7.5) containing 100 mM KCl and 5 mM MgSO₄
  • Cat. No.:
    10008-416
  • Supplier no.:
    6362-100

Specifications

About this item

>95% Pure Active Recombinant Human Neuron-Specific Enolase (NSE)

Specific activity: >8 units/ml.

Neuron-specific enolase (NSE) is a glycolytic isoenzyme which is located in central and peripheral neurons and neuroendocrine cells. This enzyme is released into the CSF when neural tissue is injured. Neoplasms derived from neural or neuroendocrine tissue may release NSE into the blood. NSE is a useful substance that has been detected in patients with certain tumors, namely - neuroblastoma, small cell lung cancer, medullary thyroid cancer, carcinoid tumors, pancreatic endocrine tumors, and melanoma. NSE shows neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons and binds in a calcium-dependent manner to cultured neocortical neurons promoting cell survival.