Filter Insertion Attachments for ASE® 100/200/300, Restek
Catalog # 10015-290
Supplier: Restek
Specifications
- Description:Filter Insertion Attachments (1 mL, 5 mL, 11 mL, 33 mL) for ASE 100/200/300
- Cat. No.:10015-290
- Supplier no.:26183
Specifications
About this item
For use with ASE® 100/200/300 extraction cells.
- Compatible with various models of ASE® extraction cells (100/200/300)
ASE 100/300 models are for use with 10 ml, 34 ml, 66 ml, and 100 ml sizes cell bodies.
ASE 200 model is for use with 5 ml, 11 ml, 22 ml, and 33 ml size cell bodies.