Cell Proliferation Assay Kit Fluorometric, BioVision
10013-674
: Avantor
:
- Storage Conditions:-20°C
- Species Reactivity:Mammalian samples
- Sample Type:Adherent and suspension cells
- Shelf Life:12 Months
- Cat. No.:10013-674
- No. of assays/kit:1000 assays
- Supplier no.:K307-1000
Non-radioactive Assay, detects as less as 25-60,000 cells
- Fluorescence (Ex/Em = 480/538 nm)
- Measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients
- Analysis of cytotoxic and cytostatic compounds such as anticancer drugs, toxic agents, and other pharmaceuticals
- Assessment of physiological mediators and antibodies that inhibit cell growth
BioVision’s Cell Proliferation Assay Kit (Fluorometric) provides an easy and quantitative method to measure cell proliferation and cytotoxicity. The assay is based on a nuclear dye that specifically binds to nucleic acid in the cell and generates green fluorescence. The generated fluorescent intensity is directly proportional to the cell number, which can be quantified by measuring fluorescence (Ex/Em = 480/538 nm). This assay kit provides a just add-and-read, non-radioactive, and high-throughput method for cell proliferation and apoptosis. The assay is rapid and more sensitive than MTT, XTT, or MTS-based assays and can detect a wide linear range of 25-60,000 cells.