About this item
>90% Pure Recombinant active human EBI-3/IL-27.
Assay data for human recombinant EBI-3 is based upon qualitative binding to anti-EBI-3 antibody.
Epstein-Barr Virus Induced Gene-3 (EBI-3), is a secreted glycoprotein belonging to the hematopoietin receptor family and related to the p40 subunit of IL-12. It was identified by its induced expression in B-lymphocytes in response to Epstein-Barr virus infection. EBI-3 forms heterodimers with p28 to form IL-27 and with p35 to form IL-35. Both IL-27 and IL-35 have anti-inflammatory and regulatory activity. Recombinant Human EBI is a non-glycosylated polypeptide chain consisting of 209 amino acids with a molecular weight of 23.3 kDa
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: