Orders LinkContactUsLinkComponent
Polypropylene In-Line Filter Holders, Advantec MFS
Polypropylene In-Line Filter Holders, Advantec MFS
Marketsource
Supplier:   Advantec MFS
Polypropylene In-Line Filter Holders, Advantec MFS
Supplier:  Advantec MFS
Marketsource
Certificates
Polypropylene In-Line Filter Holders, Advantec MFS
2 Options Available Below
Product Details & Documents
These filter holders are made from a thermoplastic polymer Polypropylene (PP) lending durability and compatibility with a wide range of chemical solvents and temperatures
Body and support screen material is PP; units come with a standard silicone o-rings.
Applications: Ultraclean and sterilize liquids or for aseptic sampling of liquids or gases at point-of-use. Use in the field or laboratory.
Exterior locking ring design allows unit to be assembled quickly and efficiently without tearing the membrane or removing from lines. Use hand tightening only.
Ordering information: Replacement parts and alternate o-ring materials are available in Accessory tab. Filters not included.
Specifications for Product Family
Specifications
  • Catalog No:
  • 101105-552
  • 101102-864
  • Max. Operating Temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
Specifications
Product Family Options
Product Information
  • Model
    Filter diameter
    Outlet connection
    Filtration area
    Inlet connection
    Max. operating pressure
    Availability
    Price
  • PP25
    25 mm
    Luer slip
    3.5 cm2
    Luer-lock
    42 psi
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $116.23
    $116.23
    Pack of 6
       $116.23
    Minimum order quantity is 1
    Supplier Number: 43303010
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101105-552
    Max. Operating Temperature
    80 °C (176 °F)
    Marketsource
  • PP47
    47 mm
    Combination 1/4" NPTM to female slip. Adaptor: 11mm O.D. hose barb included
    13.5 cm2
    Combination ¼" NPTM to female slip
    71 psi
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $73.23
    $73.23
    Pack of 1
       $73.23
    Minimum order quantity is 1
    Supplier Number: 43303020
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101102-864
    Max. Operating Temperature
    80 °C (176 °F)
    Marketsource
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • FPM locking ring O-ring
    FPM locking ring O-ring
    101105-178
    Log in for availability
    Pack of 1 - $10.22
Product Information
  • Catalog #
    Availability
    Price
  • Silicone locking ring O-ring
    Silicone locking ring O-ring
    102968-904
    Log in for availability
    Each - $7.00
Product Information
  • Catalog #
    Availability
    Price
  • EPR locking ring O-ring
    EPR locking ring O-ring
    102968-906
    Log in for availability
    Each - $5.14
Product Information
  • Catalog #
    Availability
    Price
  • Support screen
    Support screen
    102968-910
    Log in for availability
    Pack of 4 - $53.97
Product Information
  • Catalog #
    Availability
    Price
  • PP25 Inlet O-ring, Silicone
    PP25 Inlet O-ring, Silicone
    102968-912
    Log in for availability
    Pack of 12 - $64.43
Product Information
  • Catalog #
    Availability
    Price
  • PP25 Inlet O-ring, FPM
    PP25 Inlet O-ring, FPM
    102968-914
    Log in for availability
    Pack of 12 - $81.88
Product Information
  • Catalog #
    Availability
    Price
  • PP25 Inlet O-ring, EPR
    PP25 Inlet O-ring, EPR
    102968-916
    Log in for availability
    Each - $4.85
Product Information
  • Catalog #
    Availability
    Price
  • PP47 Outlet O-ring, Silicone
    PP47 Outlet O-ring, Silicone
    102968-920
    Log in for availability
    Each - $8.25
Product Information
  • Catalog #
    Availability
    Price
  • PP47 Outlet Adaptor
    PP47 Outlet Adaptor
    102968-926
    Log in for availability
    Pack of 15 - $94.91
Product Information
  • Catalog #
    Availability
    Price
  • PP47 Outlet O-ring, EPR
    PP47 Outlet O-ring, EPR
    102968-924
    Log in for availability
    Each - $8.65
Product Information
  • Catalog #
    Availability
    Price
  • PP47 Outlet O-ring, FPM
    PP47 Outlet O-ring, FPM
    102968-922
    Log in for availability
    Each - $9.25