To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Lebsche Sternum Chisel
- Length:25.4 cm (10")
- Material:Stainless steel
- Environmentally Preferable:
- Cat. No.:10197-516
- Supplier no.:55-2010
Sklar's Lebsche Sternum Chisel is used in procedures that require splitting of the sternum.
Often a mallet is used in conjunction with the chisel so that more force can be applied in the process. Splitting the sternum may be required in planned cardiovascular or thoracic procedures, or in emergency situations.