ContactUsLinkComponent
D'errico Enlarging Burr, OR Grade, Sklar
D'errico Enlarging Burr, OR Grade, Sklar
Catalog # 10185-366
Supplier:  Sklar
undefined
D'errico Enlarging Burr, OR Grade, Sklar
Catalog # 10185-366
Supplier:  Sklar
Supplier Number:  47-2476
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    D'errico Enlarging Burr
  • Material:
    Stainless steel
  • Tip configuration:
    Sharp Drill
  • Curvature:
    Straight
  • Cat. No.:
    10185-366
  • Sterility:
    Non sterile
  • Supplier no.:
    47-2476
  • Diameter:
    1.6 cm
Specifications
About this item
D'errico Enlarging Burrs are industry standard and constructed of stainless steel.
  • Reusable and latex free