About this item
The Sklar Alexander mastoid chisel is similar to the Alexander mastoid gouge but does not have the scoop-style end.
The chisel is a solid piece of stainless steel with one blunt end for hammering and a sharp, chiseled tip for removing bone at the opposite end.