Specifications
- Description:Littler Plastic Scissors
- Length:11.4 cm (4½")
- Material:Stainless steel
- Curvature:Curved
- Cat. No.:10195-212
- Supplier no.:22-2645
- Blade type:Smooth
Specifications
About this item
Sklar's Littler Suture Carrying Scissors are finger ring scissors that feature a single tiny hole in each blade through which suture material is passed and carried around the surgical site
The blunt/blunt tips may also be used for dissecting fine tissue. This product is curved with smooth edges.