Specifications
- Description:SatPax® 1200-R
- Cat. No.:94024-546
- Packaging:J-Fold, Stacked
- W×L:22.9×22.9 cm (9×9)"
- Solvent:70% isopropyl alcohol/30% deionized water
- Supplier no.:SPX120R00112
Specifications
About this item
SatPax® 1200-R combines SuperPolx® 1200 cleanroom laundered wipers composed of continuous filament polyester knit fabric with a high saturation level of 70% IPA and 30% DI water.
- Very low particles, fibers, ions, and extractables
- High abrasion resistance
- Reduces alcohol usage and preparation/handling costs
- Increases cleaning efficiency
- Smooth and durable with good wet strength
- Increases cleaning protocol consistency
This pre-wetted format provides a solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper. A high solution saturation level of 60% is suitable for regulated applications or applications requiring higher solution levels delivered to the surface.
Laundered and packaged in ISO class 4 (class 10) cleanroom. Available sterile for aseptic processing. Recommended for use in ISO class 4 (class 10) cleanroom applications and above. Reduce storage requirements and usage quantities. Eliminate squirt bottles and uncontrolled saturation levels Reduces VOC (Volatile Organic Compound) emissions. Available in a flat pack, or J-fold configurations.
Designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.