ContactUsLinkComponent
TC Adson Tissue Forceps, OR Grade, Sklar
TC Adson Tissue Forceps, OR Grade, Sklar
  89025-096
 :  Sklar
undefined
TC Adson Tissue Forceps, OR Grade, Sklar
  89025-096
 :  Sklar
 :  51-3049
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Length:
    121 mm (4¾")
  • Material:
    Tungsten carbide/stainless steel
  • Tip configuration:
    1 x 2 Teeth
  • Cat. No.:
    89025-096
  • Jaw type:
    Cross-serrated
  • Tip style:
    Straight
  • Supplier no.:
    51-3049
TC Adson Tissue Forceps are for holding and manipulating delicate tissues.
Sklar permanently bonds tungsten carbide inserts to the instrument, which provides maximum control and precision for the user, as well as increasing the length of time the instrument will remain sharp. These forceps have a wide, flat thumb grasp area that is commonly slightly grooved for an improved grip. The jaws are short and the tips are narrow. These TC Adson tissue forceps are identified by gold handles.