Reveal® G4 Rapid HIV-1/2 Antibody Test uses MedMira Rapid Vertical Flow Technology to deliver instant results in three easy steps
- HIV-1 sensitivity: 99.8% Serum, 99.8% Plasma
- HIV-2 sensitivity: 100%
- Specificity 99.1% Serum, 98.6% Plasma
Reveal® G4 LAB S/P features a built-in reagent control line, works with serum and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2 to 30°C (35 to 85°F) and test controls (1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control) are available as an accessory.
Reveal® G4 is the next generation of rapid testing using MedMira Rapid Vertical Flow Technology, and research has demonstrated that MedMira's rapid HIV test detects early infection better than other rapid tests and closely matches the performance of much more complicated enzyme immunoassay tests.
HIV-1/2 Antibody Test Controls are designed for use with Reveal® G4 Rapid HIV-1/2 Antibody Test in customer quality assurance practices. Each set of test controls contains 1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control. Each vial is sufficient to perform five Reveal® G4 tests. HIV-1/2 Antibody Test Controls have a shelf life of 12 months, and are stored at room temperature until ready for use.
This simple test provides instant results for either reactive or non-reactive specimens 5 times faster than any lateral flow test. Primary applications for Reveal® G4 include occupational exposures, preventing mother-to-child transmission, and emergency departments.