Order Entry
Puerto Rico
ContactUsLinkComponent
HEPES Buffer, 0.2M Buffer soln., pH 7.6, Low Endotoxin, Thermo Scientific
HEPES Buffer, 0.2M Buffer soln., pH 7.6, Low Endotoxin, Thermo Scientific
Certificates
HEPES Buffer, 0.2M Buffer soln., pH 7.6, Low Endotoxin, Thermo Scientific
Certificates

Specifications

  • CAS No.:
    7365-45-9
  • MDL No.:
    MFCD00006158
  • Storage Temperature:
    2°C to 8°C
  • Physical Form:
    Liquid
  • Appearance:
    Colorless

Specifications

Related Information