Order Entry
Puerto Rico
ContactUsLinkComponent
n-Butylzinc bromide 0.5 M in THF
Flammable
Corrosive
Irritant
Health
n-Butylzinc bromide 0.5 M in THF
Catalog # AAH58121-AD
CAS Number:  
Flammable
Corrosive
Irritant
Health
n-Butylzinc bromide 0.5 M in THF
Catalog # AAH58121-AD
Supplier Number:  H58121-AD
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    50 ml
  • Cat. No.:
    AAH58121-AD
  • Supplier no.:
    H58121-AD

Specifications

About this item

CAS Number:92273-73-9
 :
Formula:C₄H₉BrZn
 :202.4 g/mol
Density:0.958
Storage Temperature:Ambient
UN:3399
ADR:"4.3",III
 :Packaged under Argon in resealable ChemSeal™ bottles.
 :GHS02
 :GHS05
 :GHS07
 :GHS08
MDL Number:MFCD00671997
ADR:4.3,III
 :Packaged under argon in resealable ChemSeal™ bottles.