Orders LinkContactUsLinkComponent
Ferguson Scissors, OR Grade, Sklar
Ferguson Scissors, Premium OR-Grade, 17.8 cm (7"), Stainless steel, Blunt/Blunt, Smooth, Angled on Flat, Finger Rings, Non sterile
Catalog # 89025-534
Supplier:   Sklar
 :  15-3107
Ferguson Scissors, OR Grade, Sklar
Catalog # 89025-534
Supplier:  Sklar
Ferguson Scissors, OR Grade, Sklar
Ferguson Scissors, OR Grade, Sklar
Ferguson Scissors, Premium OR-Grade, 17.8 cm (7"), Stainless steel, Blunt/Blunt, Smooth, Angled on Flat, Finger Rings, Non sterile
Price
$241.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ferguson Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument
Censitrac Ready products are industry standard and constructed of stainless steel.
With three available tip configurations, varying curvatures and multiple lengths, these scissors are used for multipurpose cutting and dissecting. This blunt version is used for cutting materials such as sutures, tubing, or gauze.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Documents
Specifications
  • Description:
    Ferguson Scissors, Premium OR-Grade
  • Length:
    17.8 cm (7")
  • Material:
    Stainless steel
  • Curvature:
    Angled on Flat
  • Sterility:
    Non sterile
  • Blade type:
    Blunt/Blunt, Smooth
  • Handle:
    Finger Rings