Order Entry
United States
ContactUsLinkComponent
Current Balance
Current Balance
Catalog # 470219-588
undefined
Current Balance
Catalog # 470219-588
Supplier Number:  PH50255/1

Specifications

  • Description:
    Current Balance
  • Environmentally Preferable:
  • Cat. No.:
    470219-588
  • Supplier no.:
    PH50255/1

Specifications

About this item

The Current Balance is an excellent tool for studying electric currents.

  • Current Balance Measures the Force a Conducting Loop Experiences
  • For Use with the Solenoid
  • For Studying Electric Current

It's used as an accessory to the Solenoid.

Together, they measure the force experienced by a conducting loop carrying an electric current when it is placed in a magnetic field.