Order Entry
Puerto Rico
ContactUsLinkComponent
Stevens Tenotomy Hook, OR Grade, Sklar®
Stevens Tenotomy Hook, OR Grade, Sklar®
Catalog # 10197-346
Supplier:  Sklar
undefined
Stevens Tenotomy Hook, OR Grade, Sklar®
Catalog # 10197-346
Supplier:  Sklar
Supplier Number:  65-3191

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Stevens Tenotomy Hook
  • Length:
    12.1 cm (4¾")
  • Tip configuration:
    Blunt
  • Tip dimensions:
    4 mm
  • Curvature:
    Angled
  • Cat. No.:
    10197-346
  • Supplier no.:
    65-3191

Specifications

About this item

This is often used for retracting and holding ocular muscles away from the surgical site during strabismus procedures.

  • Made of stainless steel