Anti-MAP2K1 Rabbit Polyclonal Antibody
220022-186
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase Kinase 1
- Antigen Symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Form:100 μL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.
- Gene ID:5604
- Antigen Synonyms:MEK1|MAPKK1|CFC3|PRKMK1|MKK1
- Modification:phospho-Thr386
- Storage Temperature:For long-term storage, ? –20 °C is recommended. Product is stable at ? –20 °C for at least 1 year.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the human phospho-T386 of MEK1
- Purification:Antigen Affinity-purified
- Cat. No.:220022-186
- Supplier no.:PPS076
Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human