Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Dopamine beta-Hydroxylase
- Antigen Symbol:DBH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Primates,Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Epitope:C-Terminal
- Form:100 μL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and 0.09% sodium azide.
- Gene ID:1621
- Antigen Synonyms:DBM
- Storage Temperature:For long-term storage, ? –20 °C is recommended. Product is stable at ? –20 °C for at least 1 year.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Dopamine beta-Hydroxylase
- Purification:Antigen Affinity-purified
- Cat. No.:220022-176
- Supplier no.:PPS066
Specifications
About this item
Type: Primary
Antigen: DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Primate