To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Synapsin 1
- Antigen Symbol:SYN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Form:100 μL in 10 mM HEPES (pH
7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol. - Gene ID:20964
- Antigen Synonyms:SYN1a|Synapsin-1|Synapsin I|SYN1|Brain protein 4.1|SYNI|SYN1b
- Modification:phospho-Ser603
- Storage Temperature:For long-term storage, ≤ ‑20° C
is recommended. Product is stable at ≤ ‑20° C for at least 1 year. - Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-S603 of Synapsin I
- Purification:Antigen Affinity-purified
- Cat. No.:220022-148
- Supplier no.:PPS036
Type: Primary
Antigen Symbol: SYN1
Antigen Name: synapsin I
Antigen Synonyms: Syn-1, Syn1-S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Zebrafish