To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor, Metabotropic 5
- Antigen Symbol:GRM5
- Clonality:Monoclonal
- Clone:464823
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:2915
- Antigen Synonyms:GPRC1E|MGLUR5|GRM5|Metabotropic glutamate receptor 5 [Precursor]|mGluR5
- Amino Acid Number:Ser19 to Ser509
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human mGluR5 Ser19-Ser509; Accession # P41594
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220020-924
- Supplier no.:MAB45141
Specifications
About this item
Type: Primary
Antigen Symbol: GRM5
Antigen Name: glutamate receptor, metabotropic 5
Antigen Synonyms: GPRC1E, MGLUR5, PPP1R86, mGlu5
Clonality: Monoclonal
Clone: 464823
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human