Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:ENTH Domain Containing 1
- Antigen Symbol:ENTHD1
- Clonality:Monoclonal
- Clone:498403
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:953
- Antigen Synonyms:NTPDase-1|CD39|ATPDase|SPG64
- Amino Acid Number:Thr38 to Val478
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:Chinese hamster ovary cell line CHO-derived recombinant human CD39/ENTPD1 Thr38-Val478; Accession # P49961
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220020-877
- Supplier no.:MAB4397
Specifications
About this item
Type: Primary
Antigen: ENTHD1
Clonality: Monoclonal
Clone: 498403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human