Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SLAM Rat Monoclonal Antibody [clone: 459911]
Anti-SLAM Rat Monoclonal Antibody [clone: 459911]
Catalog # 220020-852
Supplier:  R&D Systems
Anti-SLAM Rat Monoclonal Antibody [clone: 459911]
Catalog # 220020-852
Supplier:  R&D Systems
Supplier Number:  MAB4330
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Signaling Lymphocytic Activation Molecule
  • Antigen Symbol:
    SLAM
  • Clonality:
    Monoclonal
  • Clone:
    459911
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Antigen Synonyms:
    CD150|CDw150|IPO-3|signaling lymphocytic activation molecule|SLAMF1|SLAM
  • Amino Acid Number:
    Thr25 to Pro242
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150 Thr25-Pro242; Accession # Q9QUM4
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-852
  • Supplier no.:
    MAB4330

Specifications

About this item

Type: Primary
Antigen: SLAM
Clonality: Monoclonal
Clone: 459911
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse